Learn More
PIP™ Bolle's Safety Spirit Protective Eyewear
Manufacturer:  Protective Industrial Products, Inc. 253ST40077
 View more versions of this product
Catalog No. 19-036-846
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yellow
One pair/pk.
Polycarbonate
Polycarbonate
Black, matte
Anti-fog,Scratch-resistant
ANSI Z87.1-2010
Soft pouch and adjustable cord
23g