PIP™ Bolle's Safety Tracker Protective Eyewear
8 base high contour polycarbonate lens with 180 degree distortion free viewing
Manufacturer: Protective Industrial Products, Inc. 253TR40088
 View more versions of this product
Catalog No. 19-036-857
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Weight (Metric) 52g
    Frame Color Black; Gray
    Lens Coating Anti-scratch; Anti-fog
    Certifications/Compliance ANSI Z87.1-2010
    Frame Material Polycarbonate; TPE
    Includes Soft pouch and adjustable cord
    Lens Material Polycarbonate
    Lens Tint Twilight
  • Description & Specifications
Related Products
Product Content Feedback