Learn More
PIP™ Bolle's Safety Tracker Protective Eyewear
8 base high contour polycarbonate lens with 180 degree distortion free viewing
Manufacturer:  Protective Industrial Products, Inc. 253TR40085
 View more versions of this product
Catalog No. 19-036-854
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Clear
ANSI Z87.1-2010
Polycarbonate
Anti-fog,Scratch-resistant
Soft pouch and adjustable cord
52g