Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Brown Jersey Gloves

Excellent for cold applications

$23.30 - $25.75

Specifications

Color Brown
Material Cotton
For Use With (Application) light protection in warehousing, farming and automotive applications
Glove Cuff Style Knit
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
19-046-388
View Documents
PIP™
95809
10.2 in. Men's 5.1 in. Dozen of 12 for $25.75
Case of 25 DZ for $586.00
Add to cart
 
19-165-219
View Documents
PIP™
95809C
9.3 in. Women's 4.3 in. Dozen of 12 for $23.30
Case of 25 DZ for $546.50
Add to cart
 
Description

Description

Ideal for light protection in warehousing, farming and the automotive industry.

  • Constructed of heavy-weight cotton jersey
  • Clute pattern
  • Knitwrist
  • Brown

Specifications

Specifications

Brown
Cotton
light protection in warehousing, farming and automotive applications
Knit
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit