Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ChemGrip™ Neoprene-Coated Gloves

Offer superior durability and protection from a wide range of chemicals such as acids, caustics, solvents and oils

$78.00 - $175.00

Specifications

For Use With (Application) refining, galvanizing, degreasing, plating, Petrochemical industries and kitchen cleaning and maintenance
Color Black
Material Neoprene
Powdered Powder-free
Size Large
View More Specs

Products 4
Catalog Number Mfr. No. Exterior Finish Length (English) Lining Price Quantity & Availability  
Catalog Number Mfr. No. Exterior Finish Length (English) Lining Price Quantity & Availability  
19-130-5317
View Documents
PIP™
578645R/L
Etched rough 18 in. Interlock knit
Dozen of 12 for $175.00
Case of 3 DZ for $525.00
Add to cart
 
19-148-383
View Documents
PIP™
578630
Smooth 12 in. Two-piece interlock Jersey knit
Dozen of 12 for $116.00
Case of 6 DZ for $696.00
Add to cart
 
19-148-382
View Documents
PIP™
578615
Smooth 9.5 in. Jersey
Dozen of 12 for $78.00
Case of 6 DZ for $468.00
Add to cart
 
19-148-381
View Documents
PIP™
578640R
Etched rough 14 in. Interlock knit
Case of 72 for $132.00
Add to cart
 
Description

Description

Ideal for refining, galvanizing, degreasing, plating, petrochemical industries and kitchen cleaning and maintenance.

  • Smooth finish gloves are available with knitwrist or 12 in. (30cm) gauntlet
  • Multi-dipped rough finish gauntlets offer excellent grip and cut resistance; available in 12 or 14 in. (30 or 36cm) lengths
  • Etched rough finish style has excellent non-slip grip and cut resistance
  • Multi-dip process provides fully liquid-proof protection
  • Fully coated; interlock or jersey lined

Specifications

Specifications

refining, galvanizing, degreasing, plating, Petrochemical industries and kitchen cleaning and maintenance
Neoprene
Large
Black
Powder-free
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit