Learn More
PIP™ CleanTeam ™ CE Single Use Latex Gloves
Used in biotechnology and medical device manufacturing.
Manufacturer:  PIP™ 100333000/XL
 View more versions of this product
Catalog No. 19-121-730D
Add to cart
Description
Description
  • Exceed standards required for use in class 100 and other controlled environments
  • Finger textured provides a good grip in wet or dry environments
  • Ambidextrous for quick and easy donning
  • Powder-Free to reduce allergic reactions
  • Beaded cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Chlorinated to harden and cleanse the surface of the glove
  • Electrostatic Dissipative (ESD) for improved performance in electronics assembly
  • 100% Silicone free
Specifications
Specifications
Textured
Nitrile
5 mil
30.5cm
Beaded/Rolled
X-Large
12 in.