Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Cotton Lisle Inspection Gloves Get IT Item

Ideal for inspection and electronics assembly or as liners under disposable and unsupported, liquid-proof gloves

Manufacturer:  Protective Industrial Products, Inc. 97501H

 View more versions of this product

Catalog No. 19-121-388



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

8.7 x 3.7 in. (22 x 9.5cm)
Economy
Cotton
Light weight
9.5cm
8.7 in.
Overcast hem
inspection and electronics assembly
Women's
3.7 in.
White
22cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit