Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Cotton Lisle Inspection Gloves

Ideal for inspection and electronics assembly or as liners under disposable and unsupported, liquid-proof gloves

Manufacturer:  PIP™ 97500H

 View more versions of this product

Catalog No. 19-044353



Add to cart

Description

Description

 

  • Constructed in a two-piece, reversible design
  • Minimize contamination of products from hands
  • 100% cotton
  • Length: 9 in. (23cm)

 

Specifications

Specifications

Safety
Overcast hem
Cotton
Light weight
10.5 cm
8.9 in.
22.5 x 10.5 cm (8.9 x 4.1 in.)
inspection and electronics assembly
Men's
4.1 in.
White
22.5 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit