Learn More
PIP™ CleanTeam™ Economy Nylon Inspection Gloves
Manufacturer:  PIP™ 98721S/PRC
 View more versions of this product
Catalog No. 11-393-1A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Manufacturer-suggested ISO Class 7 (Class 10,000)
Economical choice for hand protection or as a glove liner. Made of low-lint 40-denier tricot fabric that absorbs perspiration for a comfortable fit. Ambidextrous. Available in two-piece pattern with set-in thumb or full-fashion pattern with set-in thumb and fourchettes for tailored fit. White with color-coded hem for quick size identification. 9 in. long (23cm).
Order Info
Pack of 12 pairs.
Specifications
Specifications
Safety
Nylon
Small