Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Nylon Stretch Inspection Gloves

Ideal for inspection and electronics assembly

Manufacturer:  Protective Industrial Products, Inc. 98702

 View more versions of this product

Catalog No. 19-120-801



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

8 .9 x 3.9 in. (22.5 x 10cm)
inspection and electronics assembly, can also be used as liners under disposable and unsupported liquid proof gloves
Men's
10cm
8.9 in.
Zig zag stitched
Nylon
3.9 in.
Blue
22.5cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit