Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Cotton Canvas Gold Chore Gloves

Economy Grade, multi-purpose glove used for light protection in warehousing, farming and automotive applications.

Manufacturer:  PIP™ 93568

 View more versions of this product

Catalog No. 19-063451



Add to cart

Description

Description

  • Cotton construction keeps hands cool and protected
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Nap-out finish offers moderate heat resistance
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work

Specifications

Specifications

Safety
28 x 14 cm (11 x 5.5 in.)
Knit
light protection in warehousing, farming and automotive applications
Large, Men's
12 oz.
14 cm
11 in.
Clute
Nap-out
Economy
Cotton, Polyester
340.2 g
5.5 in.
Gold
28 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit