Learn More
PIP™ Men's Cotton / Polyester Jersey Glove
Medium Weight. Used for light protection in warehousing, farming and the automotive industry.
Manufacturer:  PIP™ 95606
 View more versions of this product
Catalog No. 19-069-700
Add to cart
Description
Description
  • Cotton/Polyester construction is breathable and decreases hand fatigue
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows replacing one glove at a time, rather than pairs
  • Straight thumb increases comfort in closed-fisted work
Specifications
Specifications
Safety
Knit
Cotton
4.5 in.
Natural
27.5 cm
27.5 x 11.5 cm (10.8 x 4.5 in.)
light protection in warehousing, farming and automotive applications
Men's
11.5 cm
10.8 in.