Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Men's Cotton / Polyester Jersey Glove

Medium Weight. Used for light protection in warehousing, farming and the automotive industry.

Manufacturer:  PIP™ 95806

 View more versions of this product

Catalog No. 19-100-569



Add to cart

Description

Description

  • Cotton/Polyester construction is breathable and decreases hand fatigue
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work
  • Brown color hides the dirt and extends the wear

Specifications

Specifications

25 x 12.5 cm (9.8 x 4.9 in.)
light protection in warehousing, farming and automotive applications
Men's
12.5 cm
9.8 in.
Knit
Cotton, Polyester
4.9 in.
Brown
25 cm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit