Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Cowhide Work Gloves

Consistent grading and superior workmanship

Manufacturer:  PIP™ 77269/XL

Catalog No. 19-161-221D


Add to cart

Description

Description

Used in cold weather applications including farming, construction, machine operation and refrigeration.

  • Premium grade all-leather top grain cowhide construction
  • Keystone thumb for superior fit and comfort
  • Thinsulate™ lining, for superior insulation without the bulk of fleece
  • Elastic back
  • Cotton color binding denotes sizes

Specifications

Specifications

Natural
Stapled
Slip-on
10.2 in.
Thinsulate™
X-Large
13.5 cm
26 x 13.5 cm (10.2 x 5.3 in.)
cold weather applications including farming, construction, machine operation and refrigeration
Blue
26 cm
Cowhide Leather
5.3 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit