Learn More
PIP™ Cut-resistant Dyneema™ Sleeves
Ideal for food processing, electronics manufacturing, glass cutting, paper processing
Manufacturer:  PIP™ 20D20
Catalog No. 19-810-612
Add to cart
Description
Description
Offers arm cut protection for applications that require large object handling and reach.
  • 100% white Dyneema polyethylene fibers up to 15 times stronger than steel, more durable than polyester and 40% stronger than Aramid fiber
  • Suitable for food contact
  • Seamless construction with knit cuff
  • Color coded hems for easy size identification
  • Washable in mild detergent; hand dry
Compliance
Meets Method ASTM F 1790-04 for cut resistance
Specifications
Specifications
Knit
1-ply
20 x 4.9 in. (50.8 x 12.5cm)
Dyneema™ Polyethylene Fiber
12.5cm
Food processing, electronics manufacturing, Glass cutting, paper processing, light duty applications
White
20 in.
50.8cm
4.9 in.
Seamless