Learn More
PIP™ Dyneema™ Cut-Resistant Polyethylene Gloves: Polyurethane-coated
Manufacturer:  PIP™ 19D123/XL
 View more versions of this product
Catalog No. 19-121-384D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Available in light and medium weight
  • White with color-coded hem
Specifications
Specifications
PE (Polyethylene)
X-Large
9 in.
23cm