Learn More
PIP Economical Chemical Goggles
Four indirect vents help minimize fogging while keeping chemicals out.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
S63094
![]() |
PIP
994401 |
N/A | Fogless replacement lens |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
S63093
![]() |
PIP
4401400/EMD |
N/A | Fogless lens goggles |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Polycarbonate lenses provide both impact and chemical splash protection
- Fit comfortably over most prescription glasses Flexible vinyl nose bridge adds comfort
Compliance
Meet ANSI Z87.1-2003 standards.