Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Economy Grade Crinkle Latex Coated Seamless Knit Cotton/Polyester Gloves

Click to view available options
Small
This item is not returnable. View return policy
This item is not returnable. View return policy

Latex crinkle coating features creases or wrinkles on the surface, designed to channel fluids away for better contact on dry or wet surfaces.

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Crinkle coating features creases or wrinkles on the surface, designed to channel fluids away for better contact on dry or wet surfaces
  • Latex is flexible, tough, and durable due to its high degree of resistance to snagging, puncture, and abrasion
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern
ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Crinkled Latex
Color Gray, Blue
Exterior Finish Fully Textured
Grade Economy
Hand Specific
Material Cotton/Polyester Blend
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 2142
For Use With (Application) Abrasive Parts/Materials, Demolition, Masonry, and Metal Fastening
Glove Cuff Style Continuous Knit
Series 39-1310
Yarn Gauge 10
Length (English) 9.3 in.
Length (Metric) 23.7 cm
Show More Show Less