Learn More
PIP™ G-Tek™ CR Plus Dyneema™ Gloves with Polyurethane Grip
Thin and flexible protection
Manufacturer:  Protective Industrial Products, Inc. 19D625/S
 View more versions of this product
Catalog No. 19-151-259
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Palm, finger
Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
Continuous knit
electronics, Glass cutting, Metal handling, food processing and meat processing
Dyneema™, Nylon, Lycra™
3.5 in.
White
23cm
Level 3
9.1 x 3.5 in. (23 x 9cm)
Red
Polyurethane
Small
9cm
9.1 in.