Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Plus Gloves

Catalog Number p-3827644
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medium weight, coated gloves made with Dyneema™

Polyethylene fibers 10 to 15 times stronger than steel, more durable than polyester and 40% stronger than aramid fiber. Used for electronics, glass cutting, metal handling, food processing and meat processing.

  • Gray spun Dyneema yarn knitted shell (13 gauge) with nylon/Lycra™
  • Gray polyurethane-coated palm and finger tips
  • Washable, resistant to chemicals, water and ultraviolet light
  • Seamless construction
  • Color-coded hems for easy size identification

Color Gray
Cut Resistance Rating ANSI A3
Material Dyneema™, Nylon, Lycra
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Electronics, glass cutting, metal handling, food processing and meat processing
Glove Cuff Color Brown
Glove Cuff Style Continuous Knit
Hem Color Brown
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit