Learn More
PIP™ G-Tek™ CR Ultra Dyneema™ Gloves with Nitrile Grip
Nitrile coated palm and fingertips offer excellent dry grip and will not absorb liquids into the coating
Manufacturer:  Protective Industrial Products, Inc. 19D750/S
 View more versions of this product
Catalog No. 19-151-272
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Palm, finger
Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Seamless
Red
Nitrile
Small
9cm
9.1 in.
Level 3
9.1 x 3.5 in. (23 x 9cm)
Continuous knit
electronics, Glass cutting, Metal handling, food processing and meat processing
Dyneema™
3.5 in.
Gray
23cm