Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Ultra Nitrile Coated Dyneema™ Gloves

Catalog Number 19151273
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Nitrile coating on the palm and fingertips enhances grip capabilities, with Dyneema™ fibers for strength and cut resistance.

  • Dyneema™ and nylon shell provides dexterity, tactile sensitivity and cut resistance
  • Thin and flexible with high protection
  • Seamless construction offers increased comfort and breathability
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Resistant to chemicals, water, and ultraviolet light
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Gray
Exterior Finish No Texture
Material Dyneema
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Electronics, Glass Cutting, Metal Handling, and Food Processing
Glove Cuff Style Continuous Knit
Hem Color Yellow
Series 19-D750
Yarn Gauge 13
Length (English) 9.4 in.
Length (Metric) 24 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit