PIP™ G-Tek™ Force Cotton/Polyester Gloves with Latex Crinkle Grip

Comfort and protection

Manufacturer: Protective Industrial Products, Inc. 39C1300/XS

 View more versions of this product

Catalog No. 19-151-288

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
    Exterior Finish Crinkle
    Glove Cuff Style Knit
    Glove Cuff Color Gray
    For Use With (Application) assembly, construction, inspection, material handling, refuse handling, landscaping and gardening
    Material Cotton, Polyester
    Liner Material Cotton, Polyester
    Size X-Small
    Coated Area Palm, finger
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit