Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP Smooth Nitrile Coated Seamless Knit Nylon Gloves

Seamless knit nylon shell increases comfort, finger dexterity, and breathability. Smooth nitrile coating is highly impermeable to liquids for enhanced grip capabilites in applications that require secure holding power.

$16.15 - $38.00

Specifications

Coating Material Nitrile
Color White
For Use With (Application) Manufacturing/Assembly Applications Where Abrasion, Cut and Puncture Resistance is Required
Glove Cuff Style Continuous Knit
Material Nylon
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-130-3719A
View Documents
PIP™
34225/S
Small
Dozen of 12 for $16.15
Case of 25 DZ for $403.75
Add to cart
 
19-130-3719B
View Documents
PIP™
34225/M
Medium
Dozen of 12 for $16.21
Case of 25 DZ for $405.25
Add to cart
 
19-130-3719C
View Documents
PIP™
34225/L
Large
Dozen of 12 for $38.00
Case of 25 DZ for $904.50
Add to cart
 
19-130-3719D
View Documents
PIP™
34225/XL
X-Large
Dozen of 12 for $35.00
Case of 25 DZ for $875.00
Add to cart
 
Description

Description

  • Solid dipped nitrile coating enhances grip capabilities, and is impermeable to liquids
  • Knit wrist helps to prevent dirt and debris from entering the glove
  • Breathable nylon back increases breathability and comfort
  • Ideal for wet or dry applications that require secure holding power
Specifications

Specifications

Nitrile
Manufacturing/Assembly Applications Where Abrasion, Cut and Puncture Resistance is Required
Nylon
White
Continuous Knit
34-225
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit