Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ BN Smooth Nitrile Coated Seamless Knit Nylon Gloves

Catalog Number p-3543467
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Smooth nitrile coating is highly impermeable to liquids to provide a tacky dry grip for applications that require secure holding power.

  • Seamless knit nylon shell increases comfort, finger dexterity, and breathability
  • Nitrile coating is highly impermeable to liquids
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Launderable
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color White, Blue
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 4131
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 34-C229
Yarn Gauge 13
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit