Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ G-Tek™ Economy Grade Foam Nitrile Coated Seamless Knit Nylon Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Knit wrist helps to prevent dirt and debris from entering the glove
- Foam nitrile coating is breathable and durable
- Gloves are resistant to chemicals, water, and ultraviolet light
- Ideal for part handling, equipment maintenance, and assembly
- Color coded hems for easy size identification
Specifications
Coated Area | Palm, Fingers |
Construction | Seamless Knit |
Coating Material | Nitrile Foam |
Color | White |
Exterior Finish | No Texture |
Grade | Economy |
Hand | Specific |
Material | Nylon |
Product Type | Seamless Knit Gloves |
Size | Medium |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction