Learn More
PIP™ Great White Dyneema™ and Lycra™ Gloves
Dyneema/Lycra blend material with polyurethane coated palm and fingers
Manufacturer:  Protective Industrial Products, Inc. 19D622/XS
 View more versions of this product
Catalog No. 19-100-947
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Palm, finger
Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
Seamless
Purple
Polyurethane
X-Small
8.8cm
8 in.
Level 3
8 x 3.5 in. (20.32 x 8.8cm)
Continuous knit
electronics, Glass cutting, sheet Metal handling, general duty, parts assembly and construction
Dyneema™, Nylon, Spandex™ blend
3.5 in.
Gray,White
20.32cm