Learn More
PIP™ Great White Dyneema™ and Lycra™ Gloves Get IT Item
Dyneema/Lycra blend material with polyurethane coated palm and fingers
Manufacturer:  Protective Industrial Products, Inc. 19D622/L
 View more versions of this product
Catalog No. 19-151-257
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Palm, finger
Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
Seamless
Brown
Polyurethane
Large
9.9cm
9.4 in.
Level 3
9.4 x 3.9 in. (24 x 9.9cm)
Continuous knit
electronics, Glass cutting, sheet Metal handling, general duty, parts assembly and construction
Dyneema™, Nylon, Spandex™ blend
3.9 in.
Gray,White
24cm