Learn More
PIP™ Top Grain Pigskin Leather Palm Drivers Glove with Hi-Vis Fabric Back
3M™ reflective band over the knuckle provides workers with greater visibility in high risk and low-light applications.
Manufacturer:  PIP™ 125368/L
 View more versions of this product
Catalog No. 19-160-823C
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Top Grain Pigskin leather construction provides breathability due its porous texture, becomes softer with use, and withstands moisture without stiffening
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Leather palm and finger tips for extra protection
  • Ideal for outdoor, cold general work applications, refrigerated warehousing, and highway construction
Specifications
Specifications
Hi-Vis Orange
Specific
Driver Gloves
Outdoor, Highway Construction, Construction, Utility Work, General Applications
Black
125-368
10 in.
21.3 lb. (Case)
Premium
Top Grain Pigskin Leather
Large
Open
Unlined
Keystone
25.5 cm