Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Top Grain Pigskin Leather Palm Drivers Glove with Hi-Vis Fabric Back

3M™ reflective band over the knuckle provides workers with greater visibility in high risk and low-light applications.

$169.62 - $255.00

Specifications

Color Hi-Vis Orange
Glove Cuff Style Open
Grade Premium
Material Top Grain Pigskin Leather
Series 125-368
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-160-823B
View Documents
PIP™
125368/M
Medium
Dozen of 12 for $255.00
Add to cart
 
19-160-823D
View Documents
PIP™
125368/XL
X-Large
Dozen of 12 for $169.62
Add to cart
 
19-160-823A
View Documents
PIP™
125368/S
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-160-823C
View Documents
PIP™
125368/L
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Top Grain Pigskin leather construction provides breathability due its porous texture, becomes softer with use, and withstands moisture without stiffening
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Leather palm and finger tips for extra protection
  • Ideal for outdoor, cold general work applications, refrigerated warehousing, and highway construction
Specifications

Specifications

Hi-Vis Orange
Premium
125-368
Open
Top Grain Pigskin Leather
Keystone
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit