Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ FR Treated Synthetic Leather Utility Gloves

Ideal for hot, heavy material handling, stamping, grinding, cut resistance and multitasking where welding could be part of the task.

$666.00

Specifications

Certifications/Compliance ANSI/ISEA 105-2000, ASTM F 1790-04
Color Black
Cut Resistance Rating Level 4
Material Dupont™ Kevlar™ Aramid Fiber, Synthetic Suede Leather
For Use With (Application) heavy material handling, stamping, grinding, cut resistance and multitasking where welding could be part of the task
View More Specs

Products 6
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Width (English) Price Quantity & Availability  
19-103-606
View Documents
PIP™
731703/S
9.4 in. Small 4 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
19-103-607
View Documents
PIP™
731703/M
9.8 in. Medium 4.25 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
19-103-608
View Documents
PIP™
731703/L
10.25 in. Large 4.5 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
19-103-609
View Documents
PIP™
731703/XL
10.6 in. X-Large 4.75 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
19-103-610
View Documents
PIP™
731703/XXL
10.8 in. 2X-Large 5 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
19-103-611
View Documents
PIP™
731703/XXXL
11 in. 3X-Large 5.25 in.
Dozen of 12 for $666.00
Case of 6 DZ for $3,620.00
Add to cart
 
Description

Description

  • FR treated synthetic suede leather palm and finger tips for heat protection
  • Kevlar™ lining offers comfort, enhanced cut protection and good heat dispersion
  • Stitched with Kevlar, adding strength and burn resistance
  • Fourchettes between the fingers enhance the natural contour of the glove for a better fit
  • Double palm offers comfort, insulation and heat protection
  • Cushioned knuckles for comfort
  • Reinforced thumb crotch for extended glove life, enhanced comfort, increased abrasion resistance and added durability in a high-wear area
  • Elastic closure sewn inside glove for safety and comfort
  • Extended slip-on cuff allows for easy on/off and forearm protection
  • FR treated material is designed to prevent after-flame, not designed for use in ARC flash or flash fire applications

Specifications

Specifications

ANSI/ISEA 105-2000, ASTM F 1790-04
Level 4
heavy material handling, stamping, grinding, cut resistance and multitasking where welding could be part of the task
Black
Dupont™ Kevlar™ Aramid Fiber, Synthetic Suede Leather
Slip-on, extended
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit