Learn More
PIP™ Premium Grade Hot Mill Gloves
Premium Grade. Used in forging, hot metal handling, castings, molds and other operations that require heat protection.
Manufacturer:  PIP™ 94932G
 View more versions of this product
Catalog No. 19-046-355
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Three-layer cotton canvas construction offers comfort, insulation and superior heat protection
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work
  • Nap-out finish for heat resistance
  • Knuckle strap for extra protection
  • Band Top cuff helps protect wrist area and allows for easy on and off
  • Starched gauntlet cuff for added forearm protection, allows for fast removal and is washable for longer wear
Specifications
Specifications
Safety
Canvas
Large
With Burlap