Learn More
PIP™ Jersey Lined Cotton Gloves with Latex Coating
Economical comfort and protection
Manufacturer:  PIP™ 553271
Catalog No. 19-065-330
Add to cart
Description
Description
Recommended for handling slippery or smooth objects.
  • Latex “crinkle” coating gives grip in wet/dry conditions
  • Offers abrasion resistance
  • Knit wrist helps prevent dirt/debris from entering
  • Straight thumb increases comfort in closed-fisted work
  • Jersey lining is soft, warm, flexible and stretchy
  • Clute cut provides a roomy fit
  • Sewn cotton shell
  • Men's size
  • Color: white/yellow
Specifications
Specifications
Safety
Knit
Latex
Men's
Crinkle
handling slippery, smooth objects, examples include Glass, tile and sheet Metal
Cotton Jersey
White, Yellow