Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Kut-Gard™ Seamless Knit Heavy Weight Dyneema™ Blended Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Seamless knit construction provides comfort, fit, and dexterity
- Dyneema™ blend enhances the gloves performance without sacrificing flexibility or dexterity and ensures maximum protection
- Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
- Special non-latex cuff dramatically reduces cuff blowout
- Excellent gripping properties
- Shrink resistant
- Can be washed in caustic detergents and bleach
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Dyneema is 15 times stronger than steel on a weight for weight basis - a High Performance Polyethylene (HPPE) fiber made from Ultra-High Molecular Weight Polyethylene (UHMWPE)
- Dyneema fiber is much thinner and more flexible than traditional materials, such as leather and cotton, while offering higher protection
Specifications
Color | White |
Cut Resistance Rating | ANSI A5 |
Material | Dyneema™, Stainless Steel Fiber, Polyester |
Product Type | Cut-resistant Gloves |
Size | Small |
For Use With (Application) | Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction