PIP™ Kut-Gard™ Kevlar™ Cut-Resistant Sleeves

Used for advanced manufacturing, glass operations,metal stamping, sheet metal, automotive, electrical and light heat applications

Manufacturer: Protective Industrial Products, Inc. 10KS14

 View more versions of this product

Catalog No. 19-816-920

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) Glass operations, Metal stamping, sheet Metal, automotive, food applications, electrical and light heat applications
    Length (English) 12 in.
    Length (Metric) 35.6cm
    Material Kevlar™
    Size 14
    Coating Material Seamless
    Color Yellow
    Cut Resistance Rating Level 4

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit