Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Aramid Fiber Gloves, Medium-Weight

Catalog Number 19100906
Click to view available options
Length (English):
9 in.
10 in.
10.5 in.
11 in.
Length (Metric):
22.9 cm
25.4 cm
26.7 cm
27.9 cm
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Green
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Aramid fiber gloves provide a comfortable and enduring protection against heat, cuts, lacerations and abrasions.

  • Kevlar™ aramid fiber provides high tensile strength relative to its weight and up to five times stronger than steel
  • Ideal for use in the automotive industry, light heat applications, glass operations and metal stamping
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Ambidextrous pattern
  • Can be used as gloves or glove liners
  • Color-coded cuff hems for easy size identification
Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Automotive industry, light heat applications, glass operations and metal stamping
Glove Cuff Color White
Glove Cuff Style Elasticated
Model 07-K300/S
Series Kut-Gard™ 07-K300
Yarn Gauge 7
Length (English) 9 in.
Length (Metric) 22.9 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit