Learn More
Marigold™ G26G Green Nitrile™ Chemical Resistant Gloves
Provides resistance to many solvents and oil based chemicals.
Manufacturer:  Marigold™ 50G26G/LX
Catalog No. 19-813-386
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Chemical and liquid protection gloves
  • Good abrasion resistance
  • Flexible and comfortable for good dexterity
  • Superior raised grip pattern for safety handling
  • Food approved
  • Cotton liner
  • Single chlorination