PIP™ Marigold Unsupported Latex Gloves, Flock-Lined

Flock lined glove, for use in low hazard areas

Manufacturer: Protective Industrial Products, Inc. 48G12YL

 View more versions of this product

Catalog No. 19-100-561

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications ISO9001:2000, ISO14001:2004, 21 CFR 170-199
    Exterior Finish Recessed diamond
    Glove Cuff Style Beaded
    For Use With (Application) cleaning and maintenance
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Flock
    Material Latex
    Size Large
    Thickness 17mil
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit