Learn More
PIP™ MaxiFlex™ Elite™ Seamless Knit Nylon Gloves with Micro-dotted Palm
Designed and developed to be light and fully breathable
Manufacturer:  Protective Industrial Products, Inc. 34244/L
 View more versions of this product
Catalog No. 19-153-144
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Micrfoam
Recommend washing gloves in tempered Water not exceeding 104°F or 40°C, a mild commercial laundry soap or detergent should be used., wash in a 5 to 10 min. cycle time
Continuous knit
Microfoam grip on palm, fingers, micro dotted palm
Nitrile
Level 1
Level 2
9.6cm
Blue
22cm
ANSI Level 1/EN388 Level 1
8.7 x 3.8 in. (22 x 9.6cm)
White
Handling small oily parts and components, general assembly, machining, micro-engineering, automotive components, logistics, warehousing and airport security
Nylon
Large
3.8 in.
CE, EN 388: 4121
8.7 in.