Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Antivibration Gloves

Half-finger antivibration gloves absorb impact, dampening shocks to the joints of the hand

Manufacturer:  PIP™ 122AV40/S

 View more versions of this product

Catalog No. 19-121-224A


Add to cart

Description

Description

Stylish black, leather-palmed gloves are available in two styles.

Style AV40

  • Features heavy-duty leather reinforcement of the knuckle
  • Offers wrist support for protection against repetitive motion injuries
Style AV71
  • Reinforced not just at the knuckle but across the back of the hand
  • Features a wraparound, reinforced wrist strap offering premium support and protection

Order Info

Also available is a half-finger, Lycra™ glove liner with the same shock-absorbing palm pad as the glove styles listed above
All three gloves and liners incorporate adjustable Velcro™ closures
Sold by the pair

Specifications

Specifications

ISO9000:2007
Half
Leather, Polyester
4.3 in.
Black
20.2cm
7.9 x 4.3 in. (20.2 x 10.8cm)
lifting, material handling, pneumatic and electric tools
Small
10.8cm
7.9 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit