Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
PIP™ Maximum Safety™ Antivibration Gloves
Half-finger antivibration gloves absorb impact, dampening shocks to the joints of the hand
Manufacturer:  PIP™ 122AV71/L
 View more versions of this product
Catalog No. 19-121-227C
Description
Description
Stylish black, leather-palmed gloves are available in two styles.
Style AV40
  • Features heavy-duty leather reinforcement of the knuckle
  • Offers wrist support for protection against repetitive motion injuries
Style AV71
  • Reinforced not just at the knuckle but across the back of the hand
  • Features a wraparound, reinforced wrist strap offering premium support and protection
Order Info
Also available is a half-finger, Lycra™ glove liner with the same shock-absorbing palm pad as the glove styles listed above
All three gloves and liners incorporate adjustable Velcro™ closures
Sold by the pair
Specifications
Specifications
Half
Lycra™
Black
Shock absorbing pad
Large