Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Cowskin Leather Palm Anti-Vibration Gloves

The glove's ergonomic design, with a heavy-duty leather palm/back, reduces hand fatigue while offering dexterity and durability.

Specifications

Color Black
For Use With (Application) Lifting, Material Handling, and Electric Tool Use
Glove Cuff Style Hook and Loop
Material Cowhide Leather
Product Type Anti-Vibration Gloves
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-121-224A
View Documents
PIP™
122AV40/S
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-121-224C
View Documents
PIP™
122AV40/L
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-121-224D
View Documents
PIP™
122AV40/XL
X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Palm pad insert absorbs and dampens shock to the joints of the hand
  • Cotton terry cloth back for breathability
  • Wrist support protects hands against injury from repetitive movements
  • Half-finger design greater dexterity, sensitivity, and breathability
  • Wrap around wrist support with hook & loop closure for secure fit, easy on/off
  • Launderable fabric extends glove life and reduces replacement costs
Specifications

Specifications

Black
Hook and Loop
Anti-Vibration Gloves
Lifting, Material Handling, and Electric Tool Use
Cowhide Leather
122-AV40
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit