Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Reinforced Leather Palm Lifting Gloves

Catalog Number 19121222A
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

This ergonomic glove is ideal for use in demanding applications that require specialized protection of the palm and wrist.

  • Ergonomic design reduces hand fatigue while offering dexterity and durability
  • Palm pad insert absorbs and dampens shock to the joints of the hand
  • Crocheted cotton mesh back for breathability
  • Wrist support protects hands against injury from repetitive movements
  • Half-finger design provides dexterity, sensitivity, and breathability
  • Hook & Loop closure for a secure fit and easy on/off
  • Launderable fabric extended glove life and reduces replacement costs
Back Material Cotton Mesh
Color Black
Hand Specific
Material Goatskin Leather
Product Type Lifting Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Specialized Palm and Wrist Protection
Glove Cuff Style Hook and Loop
Series 122-AV14
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit