Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Reinforced Leather Palm Lifting Gloves

This ergonomic glove is ideal for use in demanding applications that require specialized protection of the palm and wrist.

$22.30

Specifications

Color Black
For Use With (Application) Specialized Palm and Wrist Protection
Glove Cuff Style Hook and Loop
Material Goatskin Leather
Product Type Lifting Gloves
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-121-222A
View Documents
PIP™
122AV14/S
Small
Pair for $22.30
Dozen of 12 PR for $254.50
Add to cart
 
19-121-222C
View Documents
PIP™
122AV14/L
Large
Pair for $22.30
Dozen of 12 PR for $254.50
Add to cart
 
Description

Description

  • Ergonomic design reduces hand fatigue while offering dexterity and durability
  • Palm pad insert absorbs and dampens shock to the joints of the hand
  • Crocheted cotton mesh back for breathability
  • Wrist support protects hands against injury from repetitive movements
  • Half-finger design provides dexterity, sensitivity, and breathability
  • Hook & Loop closure for a secure fit and easy on/off
  • Launderable fabric extended glove life and reduces replacement costs
Specifications

Specifications

Black
Hook and Loop
Lifting Gloves
Cotton Mesh
Specialized Palm and Wrist Protection
Goatskin Leather
122-AV14
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit