Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety Mechanics Gloves

Ideal for automotive, aerospace or general industrial maintenance.

Manufacturer:  PIP™ 120MX2820/S

 View more versions of this product

Catalog No. 19-121-914A



Add to cart

Description

Description

  • Designed to protect the professional mechanic or enthusiast from heat, cold, cuts and abrasions
  • Amara™ synthetic leather palm and thumb is washable
  • Back is constructed of stretch fabric to provide snug fit for increased grip and finger control
  • Double stitched to provide added durability
  • Hook and loop closure for easy on/off and a secure fit
  • Hi-Vis coloring makes this product very easy to see
  • Launderable for extended life and to reduce replacement costs

Specifications

Specifications

ISO9000:2007
automotive, aerospace or general industrial maintenance
Small
10cm
8.4 in.
8.4 x 3.9 in. (21.4 x 10cm)
Leather, synthetic, Cotton, Polyester
3.9 in.
Black, Yellow
21.4cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit