Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP Maximum Safety Mechanics Gloves
Ideal for automotive, aerospace or general industrial maintenance.
$19.00
Specifications
Certifications/Compliance | ISO9000:2007 |
---|---|
For Use With (Application) | automotive, aerospace or general industrial maintenance |
Color | Black, Yellow |
Material | Cotton, Polyester, Synthetic Leather |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-121-914C
|
PIP
120MX2820/L |
Large |
Pair for $19.00
Case of 12 PR for $228.00 |
|
19-121-914B
|
PIP
120MX2820/M |
Medium |
Pair for $19.00
Case of 12 PR for $228.00 |
|
19-121-914A
|
PIP
120MX2820/S |
Small |
Pack of 12 for $19.00
Case of 6 PK for $114.00 |
|
19-121-914D
|
PIP
120MX2820/XL |
X-Large |
Pair for $19.00
Case of 12 PR for $228.00 |
|
Description
- Designed to protect the professional mechanic or enthusiast from heat, cold, cuts and abrasions
- Amara synthetic leather palm and thumb is washable
- Back is constructed of stretch fabric to provide snug fit for increased grip and finger control
- Double stitched to provide added durability
- Hook and loop closure for easy on/off and a secure fit
- Hi-Vis coloring makes this product very easy to see
- Launderable for extended life and to reduce replacement costs
Specifications
ISO9000:2007 | |
automotive, aerospace or general industrial maintenance | |
Black, Yellow | |
Cotton, Polyester, Synthetic Leather |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title