Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Mig Tig Top Grain Goatskin Welder's Gloves

All leather construction; no inside linings

Manufacturer:  PIP™ 754854/M

 View more versions of this product

Catalog No. 19-167-635B



Add to cart

Description

Description

  • Used for MIG and TIG welding, and automotive assembly
  • Provide superior workmanship, dexterity and light heat protection
  • Wing thumb; 4 leather cuff
  • Sewn with Kevlar™ thread to prolong glove life and protect seams from burning

Specifications

Specifications

11.6 x 4.7 in. (29.5 x 12cm)
mig tig welding and automotive assembly
Medium
12cm
Blue
29.5cm
Split
Leather, goatskin
4.7 in.
0.9 to 1mm
11.6 in.
ISO9001:2000
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit