Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Top Grain Goatskin Leather Mig Tig Welding Gloves

Catalog Number p-3849152
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Goatskin leather is ideal for applications that require tactile sensitivity, with Aramid Fiber stitching for increased strength and burn resistance.

  • Top Grain Goatskin leather is soft and abrasion-resistant
  • Stitched with Aramid Fiber for added strength and burn resistance
  • Gunn cut pattern delivers durability, wear, and comfort
  • Wing thumb increases comfort in open-handed work
  • Leather slip-on cuff allows for easy on and off
  • Recommended for applications that require tactile sensitivity
Color Gray
Material Top Grain Goatskin Leather
Size Medium
Style Gunn
For Use With (Application) Mig Tig Welding, Automotive Assembly
Glove Cuff Color Blue
Glove Cuff Style Gauntlet
Series 75-4854
Thumb Style Wing
Length (English) 11.6 in.
Length (Metric) 29.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit