Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP Mouse anti-Human, PE/Cy5.5, Clone: PIP/1571, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254451PECY55

Catalog No. N254451PEC5


Add to cart

Description

Description

PIP Monoclonal antibody specifically detects PIP in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PIP
PIP/1571
Flow Cytometry
Conjugated, Purified
PIP
Recombinant human GCDFP-15 protein fragment (aa 41-146) (exact sequence is proprietary)
Protein A or G purified
Cancer, Cellular Markers, Signal Transduction
Primary
5304.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit