Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ NOVAX™ Class 00 Natural Rubber Electrical Insulating Gloves - 11 in.
Click to view available options
Orange, 8
Description
Gloves are constructed of natural rubber to provide dielectric protection, and compliant with ANSI/ASTM D120 Standards and NFPA 70E for use around electrical hazards.
- Class 00 electrical insulating gloves constructed of natural rubber
- Flexible material allows workers to easily handle small, delicate parts
- Contoured shape is comfortable and helps to reduce hand fatigue
- Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
- Ideal for use around electrical and arc flash hazards
- Individually numbered for traceability
- Stamped with factory manufacturing date
- Silicone-free
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 00, Type I, NFPA 70E |
Applications | Industrial |
Color | Orange |
Exterior Finish | No Texture |
Material | Natural Rubber |
Size | 8 |
For Use With (Application) | Electrical Utility Work, Hybrid Automobiles, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Series | 147-00-11 |
Length (English) | 11 in. |
Show More |
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction